Tisk…

Anti-AFP Mouse Monoclonal Antibody (Biotin) [clone: C2 C3 MBS-12]

Dodavatel: Biotium
BNCB0810-500 BNCB0810-100
BNCB0810-500EA 15940 CZK
BNCB0810-500 BNCB0810-100
Anti-AFP Mouse Monoclonal Antibody (Biotin) [clone: C2 C3 MBS-12]
Protilátky
This antibody recognizes an oncofetal glycoprotein with a single chain of 70 kDa, which is identified as alpha fetoprotein (AFP). This MAb is highly specific to AFP and shows no cross-reaction with other oncofetal antigens or serum albumin. AFP is normally synthesized in the liver, intestinal tract, and yolk sac of the fetus. Antibody to AFP has been shown to be useful in detecting hepatocellular carcinomas (HCC) and germ cell neoplasms, especially yolk sac tumors.

Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: C2 C3 MBS-12
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR