Tisk…

Anti-CCND1 Mouse Monoclonal Antibody (Purified) [clone: CCND1/809]

Dodavatel: Biotium
BNUB0809-500 BNUB0809-100
BNUB0809-500EA 14540 CZK
BNUB0809-500 BNUB0809-100
Anti-CCND1 Mouse Monoclonal Antibody (Purified) [clone: CCND1/809]
Protilátky
Recognizes a protein of 36 kDa, identified as cyclin D1. Cyclin D1, one of the key cell cycle regulators, is a putative proto-oncogene overexpressed in a wide variety of human neoplasms. This antibody neutralizes the activity of cyclin D1 in vivo. About 60% of mantle cell lymphomas (MCL) contain a t(11; 14)(q13; q32) translocation resulting in over-expression of cyclin D1. This antibody is useful in identifying mantle cell lymphomas (cyclin D1 positive) from CLL/SLL and follicular lymphomas (cyclin D1 negative). Occasionally, hairy cell leukemia and plasma cell myeloma weakly express Cyclin D1.

Type: Primary
Antigen: CCND1
Clonality: Monoclonal
Clone: CCND1/809
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR