Tisk…
Anti-Phosphotyrosine Mouse Monoclonal Antibody (CF640R) [clone: PY793]
Dodavatel: Biotium
BNC400793-500 BNC400793-100
BNC400793-500EA 15940 CZK
BNC400793-500 BNC400793-100
Anti-Phosphotyrosine Mouse Monoclonal Antibody (CF640R) [clone: PY793]
Protilátky
Protein phosphorylation is a fundamental event in the regulation of a large number of intracellular processes. Phosphorylation of specific tyrosine residues is the result of activation or stimulation of their respective protein tyrosine kinases. The phosphorylated proteins can be auto-phosphorylated kinases or certain cellular protein substrates. Tyrosine-phosphorylated proteins are involved in signal transduction and in the regulation of cell proliferation. Antibody to phosphotyrosine provides an excellent tool for the detection, characterization, and purification of phosphotyrosineMore Product Information