Tisk…
Anti-KRT17 Mouse Monoclonal Antibody (CF488A) [clone: KRT17/778]
Dodavatel: Biotium
BNC880778-500 BNC880778-100
BNC880778-500EA 15940 CZK
BNC880778-500 BNC880778-100
Anti-KRT17 Mouse Monoclonal Antibody (CF488A) [clone: KRT17/778]
Protilátky
Cytokeratin 17 (CK17) is normally expressed in the basal cells of complex epithelia but not in stratified or simple epithelia. Antibody to CK17 is an excellent tool to distinguish myoepithelial cells from luminal epithelium of various glands such as mammary, sweat and salivary. CK17 is expressed in epithelial cells of various origins, such as bronchial epithelial cells and skin appendages. It may be considered as epithelial stem cell marker because CK17 Ab marks basal cell differentiation. CK17 is expressed in SCLC much higher than in LADC. Eighty-five percent of the triple negative breastMore Product Information
Objednat
Produktová specifikace
Symbol antigenu KRT17
Název antigenu Cytokeratin 17
Synonyma antigenu Keratin-17|KRT17|Keratin Type I Cytoskeletal 17|PCHC1|K17
Druh protilátky Primární
Klonovalita Monoclonal
Spojení CF® 488A
Klon Clone KRT17/778
Reaktivita Pig,Goat,Human,Bovine,Rat
Hostitel Mouse
Identifikační číslo genu 2785
Izotyp IgG2b kappa
Imunochemie Yes
ImunoFluorescence Yes
Koncentrace 0,1 mg/ml
Skladovací pufr PBS/0,1% BSA/0,05% azide