Tisk…

Anti-PRLR Mouse Monoclonal Antibody (Biotin) [clone: B6.2]

Dodavatel: Biotium
BNCB0741-500 BNCB0741-100
BNCB0741-500EA 15940 CZK
BNCB0741-500 BNCB0741-100
Anti-PRLR Mouse Monoclonal Antibody (Biotin) [clone: B6.2]
Protilátky
This antibody recognizes a protein of 70 kDa, identified as prolactin receptor. Prolactin is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. The initial step in its action is the binding to a specific membrane receptor (prolactin receptor), which belongs to the superfamily of class 1 cytokine receptors. The function of the prolactin receptor is mediated, at least in part, by two families of signaling molecules: Janus kinases and signal transducers and activators of transcription.

Type: Primary
Antigen: PRLR
Clonality: Monoclonal
Clone: B6.2
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR