Tisk…

Anti-XRCC6 (p70) & XRCC5 (p80) Mouse Monoclonal Antibody (Biotin) [clone: KU729]

Dodavatel: Biotium
BNCB0729-500 BNCB0729-100
BNCB0729-500EA 15940 CZK
BNCB0729-500 BNCB0729-100
Anti-XRCC6 (p70) & XRCC5 (p80) Mouse Monoclonal Antibody (Biotin) [clone: KU729]
Protilátky
Recognizes a dimer of two proteins of 70 kDa and ~80 kDa, identified as two subunits of Ku. This MAb recognizes a conformational epitope of p70/p80 dimer, which is destroyed during Western blotting. The p70/p80 dimer is important for function of a 460 kDa DNA-dependent protein kinase. Ku protein plays a role in cell signaling, proliferation, DNA repair, replication, transcriptional activation, and apoptosis.

Type: Primary
Antigen: XRCC6 (p70) & XRCC5 (p80)
Clonality: Monoclonal
Clone: KU729
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Primates
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR