Tisk…

Anti-KDR Rat Monoclonal Antibody (Purified) [clone: KDR721]

Dodavatel: Biotium
BNUB0721-500 BNUB0721-100
BNUB0721-500EA 14540 CZK
BNUB0721-500 BNUB0721-100
Anti-KDR Rat Monoclonal Antibody (Purified) [clone: KDR721]
Protilátky
CD309, also known as VEGFR2, KDR3, and Flk-1 (mouse), is a type I transmembrane glycoprotein. It is a member of the CSF-1/PDGF receptor family of type III tyrosine kinase receptors. Human VEGFR2 is mainly expressed by endothelial cells, embryonic tissues, and megakaryocytes. It plays an important role in the regulation of angiogenesis, vasculogenesis, and vascular permeability. The ligands of VEGFR2 include VEGF-A, VEGF-C, VEGF-D, and VEGF splice isoforms. Ligation of VEGFR2 with its ligands results in the receptor dimerization and auto-phosphorylation, stimulating endothelial cell proliferation and migration.

Type: Primary
Antigen: KDR
Clonality: Monoclonal
Clone: KDR721
Conjugation: Purified
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR