Tisk…

Anti-TYMS Mouse Monoclonal Antibody (CF488A) [clone: TS106]

Dodavatel: Biotium
BNC880714-500 BNC880714-100
BNC880714-500EA 15940 CZK
BNC880714-500 BNC880714-100
Anti-TYMS Mouse Monoclonal Antibody (CF488A) [clone: TS106]
Protilátky
This antibody recognizes a protein of 36 kDa, identified as Thymidylate Synthase (TS) (EC 2.1.1.45). TS converts deoxyuridine monophosphate (dUMP) to deoxythymidine monophosphate (dTMP), which is essential for DNA biosynthesis. TS is also a critical target for the fluoropyrimidines, an important group of antineoplastic drugs that are widely used in the treatment of solid tumors. Both 5-FU and fluorodeoxyuridine are converted in tumor cells to FdUMP which inactivates TS by formation of a ternary covalent complex in the presence of the folate cofactor 5,10-methylenetetrahydrofolate. Expression of TS protein is associated with response to 5-fluorouracil (5-FU) in human colorectal, gastric, head and neck, and breast carcinomas.

Type: Primary
Antigen: TYMS
Clonality: Monoclonal
Clone: TS106
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR