Tisk…
Anti-CDKN1B Mouse Monoclonal Antibody (CF488A) [clone: SX53G8]
Dodavatel: Biotium
BNC880669-500 BNC880669-100
BNC880669-500EA 15940 CZK
BNC880669-500 BNC880669-100
Anti-CDKN1B Mouse Monoclonal Antibody (CF488A) [clone: SX53G8]
Protilátky
This MAb recognizes a 27 kDa protein, identified as the p27Kip1, a cell cycle regulatory mitotic inhibitor. It is highly specific and shows no cross-reaction with other related mitotic inhibitors. In Western blotting of cell lysates from 7 human breast cancer cell lines (ZR75-1, ZR75-30, MCF-7, MDAMB453, T47D, CAL51, 734B), the antibody labels a single band corresponding to p27Kip1. It functions as a negative regulator of G1 progression and has been proposed to function as a possible mediator of TGF- induced G1 arrest. p27Kip1 is a candidate tumor suppressor gene. Reportedly, low p27More Product Information