Tisk…

Anti-TGFA Mouse Monoclonal Antibody (CF488A) [clone: MF9]

Dodavatel: Biotium
BNC880644-500 BNC880644-100
BNC880644-500EA 15940 CZK
BNC880644-500 BNC880644-100
Anti-TGFA Mouse Monoclonal Antibody (CF488A) [clone: MF9]
Protilátky
This antibody reacts with the C-terminus of TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin. The staining with Ab-1 is completely blocked by the peptide used for raising this antibody. TGF alpha (aa50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.

Type: Primary
Antigen: TGFA
Clonality: Monoclonal
Clone: MF9
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Cow, Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR