Tisk…
Anti-EGFR Mouse Monoclonal Antibody (CF647) [clone: B1D8]
Dodavatel: Biotium
BNC470643-500 BNC470643-100
BNC470643-500EA 15940 CZK
BNC470643-500 BNC470643-100
Anti-EGFR Mouse Monoclonal Antibody (CF647) [clone: B1D8]
Protilátky
This antibody reacts with the extracellular domain of EGFR and blocks the EGF/TGF alpha induced activation. It also blocks tumor growth in vivo. It is excellent for purification of EGFR. EGFR is type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin.
Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: B1D8
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human