Tisk…

Anti-MCAM Mouse Monoclonal Antibody (Biotin) [clone: C146/634]

Dodavatel: Biotium
BNCB0634-500 BNCB0634-100
BNCB0634-500EA 15940 CZK
BNCB0634-500 BNCB0634-100
Anti-MCAM Mouse Monoclonal Antibody (Biotin) [clone: C146/634]
Protilátky
The human CD146 gene maps to chromosome 11q23 and encodes a trans-membrane glycoprotein, also designated MCAM, MUC 18, that belongs to the immunoglobulin superfamily and functions as a Ca2 -independent cell adhesion molecule. CD146 expression is restricted to advanced primary and metastatic melanomas and to cell lines of the neuroectodermal lineage, but not normal melanocytes. CD146 is found on 80% of advanced primary human melanomas and correlates well with development of metastatic disease.

Type: Primary
Antigen: MCAM
Clonality: Monoclonal
Clone: C146/634
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR