Tisk…
Anti-VWF Mouse Monoclonal Antibody (CF594) [clone: 3E2D10]
Dodavatel: Biotium
BNC940633-500 BNC940633-100
BNC940633-500EA 15940 CZK
BNC940633-500 BNC940633-100
Anti-VWF Mouse Monoclonal Antibody (CF594) [clone: 3E2D10]
Protilátky
von Willebrand Factor (vWF) is a multimeric glycoprotein that is found in endothelial cells, plasma and platelets. It acts as a carrier protein for Factor VIII and promotes platelet adhesion and aggregation. vWF undergoes a variety of posttranslational modifications that influence the affinity and availability for Factor VIII, including cleavage of the propeptide and formation of N-terminal disulfide bonds. This antibody helps to establish the endothelial nature of some lesions of disputed histogenesis, e.g. Kaposi's sarcoma and cardiac myxoma. It is widely used for differentiating vascularMore Product Information
Objednat
Produktová specifikace
Symbol antigenu VWF
Environmentally Preferable
Název antigenu von Willebrand Factor
Synonyma antigenu F8VWF|von Willebrand Disease (vWD)|Factor VIII Related Antigen|Coagulation Factor VIII|von Willebrand Antigen 2
Druh protilátky Primární
Klonovalita Monoclonal
Spojení CF® 594
Klon Clone 3E2D10
Reaktivita Human
Hostitel Mouse
Identifikační číslo genu 440848
Izotyp IgG1 kappa
Western blot Yes
Imunochemie Yes
ImunoFluorescence Yes
Koncentrace 0,1 mg/ml
Skladovací pufr PBS/0,1% BSA/0,05% azide