Tisk…

Anti-KRT18 Mouse Monoclonal Antibody (Biotin) [clone: DE-K18]

Dodavatel: Biotium
BNCB0563-500 BNCB0563-100
BNCB0563-500EA 15940 CZK
BNCB0563-500 BNCB0563-100
Anti-KRT18 Mouse Monoclonal Antibody (Biotin) [clone: DE-K18]
Protilátky
This MAb reacts with a wide variety of simple epithelia. It does not react with stratified squamous epithelia. It reacts with epithelial tumors of the gastrointestinal tract, lung, breast, pancreas, ovary, and thyroid. Cytokeratin 18, which belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. It was reported that tissues from gastrointestinal tract are positive for both cytokeratin 8 and 18 but do not contain cytokeratin 14. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18.

Type: Primary
Antigen: KRT18
Clonality: Monoclonal
Clone: DE-K18
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Cat, Dog, Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR