Tisk…
Anti-BCL2 Mouse Monoclonal Antibody (CF640R) [clone: 124]
Dodavatel: Biotium
BNC400530-500 BNC400530-100
BNC400530-500EA 15940 CZK
BNC400530-500 BNC400530-100
Anti-BCL2 Mouse Monoclonal Antibody (CF640R) [clone: 124]
Protilátky
This antibody recognizes a protein of 25-26 kDa, identified as the bcl-2 α oncoprotein. It shows no cross-reaction with Bcl-x or Bax protein. Expression of bcl-2 α oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of bcl-2 α protein, whereas the normal or hyperplastic germinal centers are negative. Consequently, this antibody is valuable when distinguishing between reactive and neoplastic follicular proliferation in lymph node biopsies. It may also be used in distinguishing between those follicularMore Product Information
Objednat
Produktová specifikace
Symbol antigenu BCL2 BCL2
Environmentally Preferable
Název antigenu bcl-2 bcl-2
Synonyma antigenu Apoptosis regulator Bcl-2|B-cell CLL/lymphoma-2 Apoptosis regulator Bcl-2|B-cell CLL/lymphoma-2
Druh protilátky Primární Primární
Klonovalita Monoclonal Monoclonal
Spojení CF® 640R CF® 640R
Klon Clone 124 Clone 124
Reaktivita Human Human
Hostitel Mouse Mouse
Identifikační číslo genu 150749 150749
Izotyp IgG2a kappa IgG2a kappa
Western blot Yes Yes
Imunochemie Yes Yes
ImunoFluorescence Yes
Koncentrace 0,1 mg/ml 0,1 mg/ml
Skladovací pufr PBS/0,1% BSA/0,05% azide PBS/0,1% BSA/0,05% azide