Tisk…
Anti-CD7 Mouse Monoclonal Antibody (CF488A) [clone: C7/511]
Dodavatel: Biotium
BNC880511-100 BNC880511-500
BNC880511-100EA 5770 CZK
BNC880511-100 BNC880511-500
Anti-CD7 Mouse Monoclonal Antibody (CF488A) [clone: C7/511]
Protilátky
Recognizes a protein of 40 kDa, identified as CD7, a member of the immunoglobulin gene superfamily. Its N-terminal amino acids 1-107 are highly homologous to Ig kappa-L chains whereas the carboxyl-terminal region of the extracellular domain is proline-rich and has been postulated to form a stalk from which the Ig domain projects. CD7 is expressed on the majority of immature and mature T-lymphocytes, and T cell leukemia. It is also found on natural killer cells, a small subpopulation of normal B cells and on malignant B cells. Cross-linking surface CD7 positively modulates T cell and NK cellMore Product Information