Tisk…

Anti-SUMO1 Mouse Monoclonal Antibody (CF568) [clone: SM1/495]

Dodavatel: Biotium
BNC680495-500 BNC680495-100
BNC680495-500EA 15940 CZK
BNC680495-500 BNC680495-100
Anti-SUMO1 Mouse Monoclonal Antibody (CF568) [clone: SM1/495]
Protilátky
This MAb is specific to SUMO-1 and shows no cross-reaction with either SUMO-2 or SUMO-3. The small ubiquitin-related modifier (SUMO) proteins, which include SUMO-1, SUMO-2 and SUMO-3, belong to the ubiquitin-like protein family. Like ubiquitin, the SUMO proteins are synthesized as precursor proteins that undergo processing before conjugation to target proteins. Also, both utilize the E1, E2, and E3 cascade enzymes for conjugation. However, SUMO and ubiquitin differ with respect to targeting. Ubiquitination predominantly targets proteins for degradation, whereas sumoylation targets proteins to a variety of cellular processing, including nuclear transport, transcriptional regulation, apoptosis and protein stability. The unconjugated SUMO-1 protein localizes to the nuclear membrane.

Type: Primary
Antigen: SUMO1
Clonality: Monoclonal
Clone: SM1/495
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR