Tisk…

Anti-ODC1 Mouse Monoclonal Antibody (Biotin) [clone: ODC1/485]

Dodavatel: Biotium
BNCB0485-500 BNCB0485-100
BNCB0485-500EA 15940 CZK
BNCB0485-500 BNCB0485-100
Anti-ODC1 Mouse Monoclonal Antibody (Biotin) [clone: ODC1/485]
Protilátky
Recognizes a 53 kDa protein, identified as the Ornithine Decarboxylase (ODC-1). ODC is the initial and rate-limiting enzyme in the biosynthetic pathway of polyamines and is involved in the conversion of ornithine to putrescine. The biological activity of ODC-1 is rapidly induced in response to virtually all agents known to promote cell proliferation including hormones, drugs, growth factors, mitogens, and tumor promoters. Reportedly, ODC mRNA levels are elevated in lung carcinomas as well as in colon adenomas and carcinomas. ODC activity in colorectal carcinomas is greater than those in adenomas and normal mucosa.

Type: Primary
Antigen: ODC1
Clonality: Monoclonal
Clone: ODC1/485
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR