Tisk…
Anti-CD5 Mouse Monoclonal Antibody (CF405S) [clone: C5/473]
Dodavatel: Biotium
BNC040473-500 BNC040473-100
BNC040473-500EA 15940 CZK
BNC040473-500 BNC040473-100
Anti-CD5 Mouse Monoclonal Antibody (CF405S) [clone: C5/473]
Protilátky
Recognizes a 67 kDa transmembrane protein, which is identified as CD5. The CD5 antigen is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. Anti-CD5 is a pan T-cell marker that also reacts with a range of neoplastic B-cells, e.g. chronic lymphocytic leukemia/small lymphocytic lymphoma (CLL/SLL), mantle cell lymphoma, and a subset (~10%) of diffuse large B-cell lymphoma. CD5 aberrant expression is useful in making a diagnosis of mature T-cell neoplasms. Anti-CD5 detection is diagnostic in CLL/SLL within a panelMore Product Information