Tisk…
Anti-HLA-DRB1 Mouse Monoclonal Antibody (CF647) [clone: L243]
Dodavatel: Biotium
BNC470458-500 BNC470458-100
BNC470458-500EA 15940 CZK
BNC470458-500 BNC470458-100
Anti-HLA-DRB1 Mouse Monoclonal Antibody (CF647) [clone: L243]
Protilátky
This MAb reacts with the HLA-DRB1 antigen, a member of MHC class II molecules. It does not cross react with HLA-DP and HLA-DQ. It binds a conformational epitope on HLA-DR, which depends on the correct folding of the α/β heterodimer. This MAb has been reported to block mixed lymphocyte reactions. The L243 antibody recognizes a different epitope than the LN3 monoclonal antibody, and these antibodies do not cross-block binding to each other's respective epitopes. HLA-DR is a heterodimeric cell surface glycoprotein comprised of a 36kD alpha (heavy) chain and a 28kD beta (light) chain. It isMore Product Information
Objednat
Produktová specifikace
Symbol antigenu HLA-DRB1
Název antigenu HLA-DRB
Environmentally Preferable
Druh protilátky Primární
Klonovalita Monoclonal
Spojení CF® 647
Klon Clone L243
Reaktivita Baboon,Monkey,Squirrel,Chimpanzee,Human,Dog
Hostitel Mouse
Identifikační číslo genu 534322
Izotyp IgG2a kappa
ImunoFluorescence Yes
Koncentrace 0,1 mg/ml
Skladovací pufr PBS/0,1% BSA/0,05% azide