Tisk…

Anti-SILV Mouse Monoclonal Antibody (CF594) [clone: HMB45]

Dodavatel: Biotium
BNC940444-500 BNC940444-100
BNC940444-500EA 15090 CZK
BNC940444-500 BNC940444-100
Anti-SILV Mouse Monoclonal Antibody (CF594) [clone: HMB45]
Protilátky
By immunohistochemistry, this antibodyspecifically recognizes a protein in melanocytes and melanomas. This MAb reacts with junctional and blue nevus cells and variably with fetal and neonatal melanocytes. Intradermal nevi, normal adult melanocytes, and non-melanocytic cells are negative. It does not stain tumor cells of epithelial, lymphoid, glial, or mesenchymal origin. Metastatic amelanotic melanoma can often be confused with a variety of poorly differentiated carcinomas, large cell lymphomas, and sarcomas using H & E stains alone. It is also difficult to differentiate melanoma from spindle cell carcinomas and various types of mesenchymal neoplasms. This MAb stains fetal and neonatal melanocytes, junctional and blue nevus cells, and malignant melanoma. This MAb also stains Angiomyolipoma (PEComa).

Type: Primary
Antigen: SILV
Clonality: Monoclonal
Clone: HMB45
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR