Tisk…

Anti-CD6 Mouse Monoclonal Antibody (CF405S) [clone: 3F7B5]

Dodavatel: Biotium
BNC040428-500 BNC040428-100
BNC040428-500EA 15940 CZK
BNC040428-500 BNC040428-100
Anti-CD6 Mouse Monoclonal Antibody (CF405S) [clone: 3F7B5]
Protilátky
CD6 is a type I transmembrane glycoprotein that contains a 24-amino acid signal sequence, three extracellular scavenger receptor cysteine-rich (SRCR) domains, a membrane-spanning domain and a 44-amino acid cytoplasmic domain. The CD6 glycoprotein is tyrosine phosphorylated during TCR-mediated T cell activation. CD6 shows significant homology to CD5. CD6 is present on mature thymocytes, peripheral T cells and a subset of B cells. Antibodies to CD6 are used to deplete T cells from bone marrow transplants to prevent graft versus host disease.

Type: Primary
Antigen: CD6
Clonality: Monoclonal
Clone: 3F7B5
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR