Tisk…

Anti-PODXL Mouse Monoclonal Antibody (CF594) [clone: 3D3]

Dodavatel: Biotium
BNC940427-500 BNC940427-100
BNC940427-500EA 15090 CZK
BNC940427-500 BNC940427-100
Anti-PODXL Mouse Monoclonal Antibody (CF594) [clone: 3D3]
Protilátky
Podocalyxin is a member of the CD34 transmembrane sialomucin family. It is over-expressed on the podocyte foot projections and plays essential roles in kidney development and homeostasis, blood filtration and urine formation. It is also expressed on vascular endothelia, hematopoietic progenitors and a subset of neurons. Overexpression of podocalyxin may be linked to more aggressive tumor behavior. Podocalyxin antibody can identify podocytes in the urine (podocyturia) that may indicate glomerular disease, pre-eclampsia, and other kidney pathology.

Type: Primary
Antigen: PODXL
Clonality: Monoclonal
Clone: 3D3
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rabbit, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR