Tisk…

Anti-UPK3A Rabbit Polyclonal Antibody (CF488A) [clone: Rabbit PAb]

Dodavatel: Biotium
BNC880409-500 BNC880409-100
BNC880409-500EA 15940 CZK
BNC880409-500 BNC880409-100
Anti-UPK3A Rabbit Polyclonal Antibody (CF488A) [clone: Rabbit PAb]
Protilátky
Uroplakin-3A (UPK3A) is a component of the asymmetric unit membrane (AUM). It is a highly specialized bio-membrane made by terminally differentiated urothelial cells. The protein may play an important role in AUM-cytoskeleton interaction in terminally differentiated urothelial cells. UPK3A also contributes to the formation of urothelial glycocalyx, which may play an important role in preventing bacterial adherence through FimH bacterial protein binding leading to bladder infection. UPK3A has been shown as a helpful marker for the detection of bladder cancer.

Type: Primary
Antigen: UPK3A
Clonality: Polyclonal
Clone: Rabbit PAb
Conjugation: CF488A
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR