Tisk…
Anti-CD7 Mouse Monoclonal Antibody (Purified) [clone: B-F12]
Dodavatel: Biotium
BNUB0310-500 BNUB0310-100
BNUB0310-500EA 14540 CZK
BNUB0310-500 BNUB0310-100
Anti-CD7 Mouse Monoclonal Antibody (Purified) [clone: B-F12]
Protilátky
Recognizes a protein of 40 kDa, identified as CD7 (also known as gp40, Leu9). CD7 is a member of the immunoglobulin gene superfamily. Its N-terminal amino acids 1-107 are highly homologous to Ig kappa-L chains whereas the carboxyl-terminal region of the extracellular domain is proline-rich and has been postulated to form a stalk from which the Ig domain projects. CD7 is expressed on the majority of immature and mature T-lymphocytes, and T cell leukemia. It is also found on natural killer cells, a small subpopulation of normal B cells and on malignant B cells. Cross-linking surface CD7More Product Information