Tisk…
Anti-CD48 Mouse Monoclonal Antibody (CF594) [clone: 156-4H9]
Dodavatel: Biotium
BNC940307-500 BNC940307-100
BNC940307-500EA 15940 CZK
BNC940307-500 BNC940307-100
Anti-CD48 Mouse Monoclonal Antibody (CF594) [clone: 156-4H9]
Protilátky
Reacts with human CD48, a 45 kDa glycosyl phophatidyl-inositol (GPI)-anchored cell surface protein. CD48 is strongly expressed on lymphocytes and monocytes and weakly on granulocytes but is absent on platelets, fibroblasts, epithelium and endothelium. CD48 is one of marker for detecting the defects of GPI anchoring structure on the patients with paroxysmal nocturnal hemoglobulinuria (PNH) and serves as a low affinity ligand for CD2.
Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: 156-4H9
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human