Tisk…
Anti-FN1 Mouse Monoclonal Antibody (CF647) [clone: HFN7.1]
Dodavatel: Biotium
BNC470270-500 BNC470270-100
BNC470270-500EA 15940 CZK
BNC470270-500 BNC470270-100
Anti-FN1 Mouse Monoclonal Antibody (CF647) [clone: HFN7.1]
Protilátky
Fibronectin is a dimeric glycoprotein of 440 kDa, which is present in cells, extracellular matrix, and blood. It possesses at least four binding sites for collagen, glycosaminoglycans, transglutaminase, and a cell surface receptor. Fibronectin is involved in cell adhesion, tissue organization, and wound healing. This MAb is directed against the peptide core and reacts with both the plasma and cellular forms of fibronectin. It blocks the fibronectin-medicated cell attachment not by disrupting the collagen-fibronectin interaction, but by interfering with the attachment of fibronectin to itsMore Product Information
Objednat
Produktová specifikace
Symbol antigenu FN1
Název antigenu Fibronectin
Environmentally Preferable
Synonyma antigenu FN1|Ugl-Y3|FNZ|Migration stimulating factor (MSF)|GFND2|Cold insoluble globulin (CIG)|LETS|GFND|FINC
Druh protilátky Primární
Klonovalita Monoclonal
Spojení CF® 647
Klon Clone HFN7.1
Reaktivita Human
Hostitel Mouse
Identifikační číslo genu 203717
Izotyp IgG1 kappa
Imunochemie Yes
ImunoFluorescence Yes
Koncentrace 0,1 mg/ml
Skladovací pufr PBS/0,1% BSA/0,05% azide