Tisk…

Anti-ABO Mouse Monoclonal Antibody (Purified) [clone: HE-14]

Dodavatel: Biotium
BNUB0235-500 BNUB0235-100
BNUB0235-500EA 14540 CZK
BNUB0235-500 BNUB0235-100
Anti-ABO Mouse Monoclonal Antibody (Purified) [clone: HE-14]
Protilátky
This antibody recognizes human blood group A (monofucosyl and difucosyl A antigens with chain types 1, 2, 3, 4, 5, 6) and Forssmann antigen. It is also reactive with the immuno-dominant A trisaccharide. Blood group antigen expression in human colon cancer was studied by means of two monoclonal antibodies of broad anti-A (HE-14) and anti-type 3 and type 4 chain-based A and H (HE-10) specificity. These antigens were proved to re-appear in tumors of the distal colon, the HE-10 antibody reacting more frequently (9 out of 12 samples) than HE-14 (5 out of 12 samples) and frequently with supra-nuclear staining of the cytoplasm probably in those places of the Golgi apparatus where carbohydrate antigens are synthesized. This staining pattern is characteristic of HE-10 in normal colonic mucosa as well. With HE-14, staining was often absent in less differentiated tumors, while HE-10 did react in such tumors. In some cases, these two antibodies gave different staining patterns in parallel sections from the same tissue sample, primarily at the cellular level. Three out of 12 cases showed blood group antigen expression in the mucosa of the distal colon adjacent to the tumor only when HE-10 Ab was used.

Type: Primary
Antigen: ABO
Clonality: Monoclonal
Clone: HE-14
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR