Tisk…

Anti-SILV Mouse Monoclonal Antibody (Purified) [clone: NKI-beteb]

Dodavatel: Biotium
BNUB0226-500 BNUB0226-100
BNUB0226-500EA 14540 CZK
BNUB0226-500 BNUB0226-100
Anti-SILV Mouse Monoclonal Antibody (Purified) [clone: NKI-beteb]
Protilátky
By immunohistochemistry, this antibody specifically recognizes a protein in melanocytes and melanomas. This MAb reacts with junctional and blue nevus cells and variably with fetal and neonatal melanocytes. Intradermal nevi, normal adult melanocytes, and non-melanocytic cells are negative. It does not stain tumor cells of epithelial, lymphoid, glial, or mesenchymal origin.This Mab labels formalin-fixed, paraffin-embedded melanomas and other tumors showing melanocytic differentiation.

Type: Primary
Antigen: SILV
Clonality: Monoclonal
Clone: NKI-beteb
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Horse, Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR