Tisk…

Anti-CD37 Mouse Monoclonal Antibody (CF594) [clone: IPO-24]

Dodavatel: Biotium
BNC940170-500 BNC940170-100
BNC940170-500EA 15940 CZK
BNC940170-500 BNC940170-100
Anti-CD37 Mouse Monoclonal Antibody (CF594) [clone: IPO-24]
Protilátky
Recognizes a protein of 33-55 kDa, identified as CD37 (Workshop V; Code CD37.7). CD37 is strongly expressed on normal and neoplastic mature (sIg ) B-lymphocytes. In B-cell ontogeny, CD37 appears after the pre-B-cell stage, is maintained during peripheral B-cell development and is lost upon terminal differentiation into plasma cells.1 CD37 is also present, at low densities, on resting and activated T cells, neutrophils, monocytes, and some myelomonocytic leukemia cells. It is absent from platelets, erythrocytes. CD37 is a member of a family of tetraspan transmembrane proteins, including CD9, CD53, CD63, CD81, and CD82. It associates other tetraspan transmembrane proteins and MHC class II molecules to form a large complex at the surface of B cells and play a role in signal transduction. CD37 is a valuable and stable marker for peripheral mature B-cells and corresponding malignancies like B-cell chronic lymphocytic leukemia (B-CLL), hairy cell leukemia (HCL), and all types of B-cell non-Hodgkin'' lymphoma (B-NHL).

Type: Primary
Antigen: CD37
Clonality: Monoclonal
Clone: IPO-24
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR