Tisk…
Anti-CD36 Mouse Monoclonal Antibody (CF640R) [clone: 185-1G2]
Dodavatel: Biotium
BNC400169-500 BNC400169-100
BNC400169-500EA 15940 CZK
BNC400169-500 BNC400169-100
Anti-CD36 Mouse Monoclonal Antibody (CF640R) [clone: 185-1G2]
Protilátky
Recognizes a protein of 80 kDa-90 kDa, identified as CD36 (Workshop IV; Code P-26). Its epitope maps between aa155-183. It is expressed on platelets, monocytes and macrophages, microvascular endothelial cells, erythrocyte precursors, mammary epithelial cells, and some macrophage derived dendritic cells. CD36 acts as a receptor for thrombospondin (TSP), collagen types I, IV and V, P. falciparum malaria-infected erythrocytes, and sickle erythrocytes. It also functions as a scavenger receptor, mediating macrophage uptake of oxidized low-density lipoprotein (LDL) and recognition of apoptoticMore Product Information