Tisk…
Anti-S100A1 Mouse Monoclonal Antibody (CF405S) [clone: 4C4.9]
Dodavatel: Biotium
BNC040143-500 BNC040143-100
BNC040143-500EA 15940 CZK
BNC040143-500 BNC040143-100
Anti-S100A1 Mouse Monoclonal Antibody (CF405S) [clone: 4C4.9]
Protilátky
S100 belongs to the family of calcium binding proteins. S100A and S100B proteins are two members of the S100 family. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S-100 protein has been found in normal melanocytes, Langerhans cells, histiocytes, chondrocytes, lipocytes, skeletal and cardiac muscle, Schwann cells, epithelial and myoepithelial cells of the breast, salivary and sweat glands, as well as in glial cells. Neoplasms derived from these cells also express S-100 protein, albeit non-uniformly. A large number of well-differentiated tumors ofMore Product Information
Objednat
Produktová specifikace
Symbol antigenu S100A1
Název antigenu S100
Environmentally Preferable
Synonyma antigenu S100 Calcium Binding Protein A1|Bpb|S100 Alpha Chain|S100 Calcium Binding Protein B|NEF|S100-A1|S100 Beta Chain|S100 Calcium Binding Protein Beta Neural
Druh protilátky Primární
Klonovalita Monoclonal
Spojení CF® 405S
Klon Clone 4C4.9
Reaktivita Human,Bovine,Rat,Mouse
Hostitel Mouse
Identifikační číslo genu 515715
Izotyp IgG2a kappa
Western blot Yes
Imunochemie Yes
ImunoFluorescence Yes
Koncentrace 0,1 mg/ml
Skladovací pufr PBS/0,1% BSA/0,05% azide