Tisk…

Anti-Human Nuclear Antigen Mouse Monoclonal Antibody (CF647) [clone: 235-1]

Dodavatel: Biotium
BNC470099-500 BNC470099-100
BNC470099-500EA 15940 CZK
BNC470099-500 BNC470099-100
Anti-Human Nuclear Antigen Mouse Monoclonal Antibody (CF647) [clone: 235-1]
Protilátky
This MAb is an excellent marker for human cells in xenographic model research. It reacts specifically with human cells. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. MAb 235-1 recognizes an antigen associated with the nuclei in human cells. It can be used to stain the nuclei in cell or tissue preparations and can be used as a nuclear marker in subcellular fractions. It produces a speckled pattern in normal and malignant cells and may be used to stain the nuclei of cells in fixed or frozen tissue sections. It can also be used with paraformaldehyde fixed frozen tissue or cell preparations.

Type: Primary
Antigen: Human Nuclear Antigen
Clonality: Monoclonal
Clone: 235-1
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Non-human primates
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR