Tisk…

Anti-ST6GAL1 Mouse Monoclonal Antibody (Purified) [clone: LN-1]

Dodavatel: Biotium
BNUB0055-500 BNUB0055-100
BNUB0055-500EA 14540 CZK
BNUB0055-500 BNUB0055-100
Anti-ST6GAL1 Mouse Monoclonal Antibody (Purified) [clone: LN-1]
Protilátky
Recognizes a neuraminidase-sensitive sialoprotein (CDw75), present on cell membrane and cytoplasm of germinal center B-cells and derived lymphomas. This MAb reacts with RBC precursors of bone marrow, ductal and ciliated epithelial cells of kidney, breast, prostate, pancreas, lung, and with glioblastomas, astrocytomas, and Reed Sternberg cells in lymphocyte predominant Hodgkin's disease. It is shown to be a helpful antibody for ascribing a B-cell phenotype in known lymphoid tissues.

Type: Primary
Antigen: ST6GAL1
Clonality: Monoclonal
Clone: LN-1
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR