Tisk…

Anti-UCHL1 Mouse Monoclonal Antibody (CF640R) [clone: 31A3]

Dodavatel: Biotium
BNC400046-500 BNC400046-100
BNC400046-500EA 15940 CZK
BNC400046-500 BNC400046-100
Anti-UCHL1 Mouse Monoclonal Antibody (CF640R) [clone: 31A3]
Protilátky
This MAb reacts with a protein of 20-30 kDa, identified as PGP9.5, also known as ubiquitin carboxyl-terminal hydrolase-1 (UchL1). Initially, PGP9.5 expression in normal tissues was reported in neurons and neuroendocrine cells but later it was found in distal renal tubular epithelium, spermatogonia, Leydig cells, oocytes, melanocytes, prostatic secretory epithelium, ejaculatory duct cells, epididymis, mammary epithelial cells, Merkel cells, and dermal fibroblasts. Furthermore, immunostaining for PGP9.5 has been shown in a wide variety of mesenchymal neoplasms as well. A mutation in PGP9.5 gene is believed to cause a form of Parkinson's disease.

Type: Primary
Antigen: UCHL1
Clonality: Monoclonal
Clone: 31A3
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Cow, Human, Mouse, Pig, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR