Tisk…

Anti-MUC2 Mouse Monoclonal Antibody (CF594) [clone: CCP58]

Dodavatel: Biotium
BNC940032-500 BNC940032-100
BNC940032-500EA 15940 CZK
BNC940032-500 BNC940032-100
Anti-MUC2 Mouse Monoclonal Antibody (CF594) [clone: CCP58]
Protilátky
Recognizes a single glycoprotein of 520 kDa, identified as mucin 2 (MUC2). This MAb shows no cross-reaction with human milk fat globule membranes, MUC1, or MUC3. Its epitope has been defined as GTQTP (GlyThrGlnThrPro). Mucins are high molecular weight glycoproteins, which constitute the major component of the mucus layer that protects the gastric epithelium. MUC2 is specifically expressed in goblet cells of the small intestine & colon; in about 65% of colonic carcinomas and about 40% of gastric carcinomas. MUC2 is rarely expressed outside of the GI tract with the exceptions of mucinous carcinoma of breast and clear cell-type carcinomas of the ovary.

Type: Primary
Antigen: MUC2
Clonality: Monoclonal
Clone: CCP58
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR