Tisk…

Anti-PTPRC Mouse Monoclonal Antibody (CF568) [clone: UCHL-1]

Dodavatel: Biotium
BNC680003-500 BNC680003-100
BNC680003-500EA 15090 CZK
BNC680003-500 BNC680003-100
Anti-PTPRC Mouse Monoclonal Antibody (CF568) [clone: UCHL-1]
Protilátky
Recognizes a 180-185 kDa protein, identified as isoform of leukocyte common antigen (CD45RO) (4th Leucocyte Typing Workshop: Code No. N31). The epitope recognized by this antibody is sensitive to neuraminidase digestion. This antibody reacts with mature activated T-cells, most thymocytes, and a sub-population of resting T-cells within both CD4 and CD8 subsets. It shows no reactivity with normal B or natural killer cells, but reacts with granulocytes and monocytes. Reportedly, it is useful to identify T-cell lymphomas and leukemias. It rarely stains NK cells or B-cell lymphomas.

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: UCHL-1
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Chimpanzee, Human, Marmoset
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR