Tisk…
Conductivity cells, two-electrode, Orion™
Dodavatel: Thermo Orion
013016MD 011010A 011020A 011510MD 018020MD
663-0181EA 23630 CZK
663-0181 ORIO011010A 663-0185 663-0188 663-0190
Conductivity cells, two-electrode, Orion™
Elektrody Cely vodivostní
These versatile conductivity cells are intended for laboratory and field use.
  • Available in a variety of materials and measurement ranges
011010A and 011020A are designed for use with 555A, 550A, 162A, 136A, 135A, 131S, 130A meters.
011010, 011020 and 011050 are designed for use with 150Aplus, 145Aplus, 125Aplus, 115Aplus, 105Aplus meters.
011050MD, 011510MD, 013016MD and 018020MD are designed for use with Star series meters.