Tisk…

L(+)-Diamoniumtartrát, white crystalline powder

Dodavatel: MP Biomedicals

Synonyms: (R,R)-(+)-Diammonium tartrate, (R,R)-(+)-Ammonium tartrate dibasic, Ammonium tartrate dibasic, L(+)-Tartaric acid diammonium salt, (+)-Diammonium L-tartrate, (R,R)-(+)-Tartaric acid diammonium salt, L(+)-Ammonium tartrate dibasic, Ammonium L-(+)-tartrate

0219504280 0219504290
ICNA0219504280EA 1890 CZK
ICNA0219504280 ICNA0219504290
L(+)-Diamoniumtartrát, white crystalline powder
L(+)-Diamoniumtartrát

Ammonium tartrate, the ammonium salt of tartaric acid, is used in such applications as cell culture and chromatography.


Ammonium tartrate, the ammonium salt of tartaric acid, is used in such applications as cell culture and chromatography. Ammonium tartrate has been used to displace sodium for the analysis of a modified oligonucleotide by matrix-assisted laser desorption/ionization post-source decay (MALDI-PSD). Ammonium tartrate is also utilised in electron spin resonance (ESR) dosimetry research, and has been probed as a more sensitive standard than L-alanine for high dose determinations.


Soluble in water 1 M at 20 °C, clear, colourless.

Vzorec: C₄H₁₂N₂O₆
MW: 184.15
Storage Temperature: Ambient
MDL Number: MFCD00013073
CAS číslo: 3164-29-2
UN: 3077
ADR: 9 III

Objednat

Výsledky specifikačního testu

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR