Tisk…
Tetraboritan dvojsodný, anhydrous ~99%, white crystals
Dodavatel: MP Biomedicals
Nebezpečí
Synonyms: Borax, Sodium tetraborate
0219030980 0219030990 0219030991
ICNA0219030980EA 806 CZK
ICNA0219030980 ICNA0219030990 ICNA0219030991
Tetraboritan dvojsodný, anhydrous ~99%, white crystals
Tetraboritan dvojsodný
Borax anhydrous typically used as a buffer component.
The Merck Index lists other uses as: used in the manufacturing of glazes and enamels, a component in tanning agents; curing and preserving skins, a agent used in cleaning compounds, used to artificially age wood, as a preservative, either alone or with other antiseptics against wood fungus, used in fireproofing fabrics and wood, used as antiseptic, used in detergents, an astringent for mucous membranes.
Anhydrous borax is slowly soluble in water. Insoluble in ethanol.
Vzorec: Na₂B₄O₇
MW: 201.22
Bod varu: 1575
Bod tání: 742.5 °C (lit.)
MDL Number: MFCD00081185
CAS číslo: 1330-43-4
Merck Index: 12,08733
Objednat
Výsledky specifikačního testu
Identity Test Passes
Purity ~99%
Moisture Content < 1,0%
Iron as Fe <0,004%
sulphate as SO4 <0,05
B2O3 68 to 70%
Na2O 30 to 40%