Tisk…

Retinol all trans all trans (synthetic), orange powder

Dodavatel: MP Biomedicals
Varování

Synonyms: all-trans-Retinol, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-ol, Vitamin A alcohol

0219026825 0219026880 0219026883 0219026890 0219026891
ICNA0219026825EA 1650 CZK
ICNA0219026825 ICNA0219026880 ICNA0219026883 ICNA0219026890 ICNA0219026891
Retinol all trans all trans (synthetic), orange powder
Retinol all trans

Retinol is one of the animal forms of vitamin A. It is a diterpenoid and an alcohol.


Retinol is an effective antioxidant displaying lipoperoxy radical scavenging activity. The interactions between Retinol and Vitamin E (α-tocopherol) in suppressing lipid peroxidation were observed in bovine retinal membrane preparations. It may influence the production of transition vesicles by stimulating the activity of a protein disulfide isomeraselike activity involved in vesicle formation. Retinol (10 µM) stimulated DNA synthesis and possibly repair mechanisms in Sertoli cells of rat.


Biochem/physiol Actions: Ligand for both the retinoic acid receptor (RAR) and the retinoid X receptor (RXR) that act as transcription factors to regulate the growth and differentiation of normal and malignant cells.


Soluble in chloroform (50 mg/ml), absolute alcohol, methanol, ether, fats and oils; practically insoluble in water and glycerol.

Vzorec: C₂₀H₃₀O
MW: 286,46 g/mol
Bod tání: 61…63 °C (lit.)
Storage Temperature: Freezer
MDL Number: MFCD00001552
CAS číslo: 68-26-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR