Tisk…

(+/-)-Thalidomide ≥99%, powder

Dodavatel: MP Biomedicals

Synonyms: 2-(2,6-Dioxopiperidin-3-yl)isoindoline-1,3-dione, N-(2,6-Dioxo-3-piperidinyl)phthalimide

0215875350 0215875380 0215875383
ICNA0215875350EA 1680 CZK
ICNA0215875350 ICNA0215875380 ICNA0215875383
(+/-)-Thalidomide ≥99%, powder
(+/-)-Thalidomide

(±)-Thalidomide selectively inhibits biosynthesis of tumor necrosis factor α (TNF-α); inhibitor of angiogenesis; immunosuppressive; sedative; teratogen.


Soluble in DMSO (>20 mg/ml).

Vzorec: C₁₃H₁₀N₂O₄
MDL Number: MFCD00153873
CAS číslo: 50-35-1
UN: 2811
ADR: 6.1,III

Objednat

Výsledky specifikačního testu

Identity Test Passes
Purity ≥99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR