Tisk…

2,2,4,4,6,8,8-Heptamethylnonane, clear, colourless liquid

Dodavatel: MP Biomedicals
Nebezpečí

0215732225 0215732280
ICNA0215732225EA 2170 CZK
ICNA0215732225 ICNA0215732280
2,2,4,4,6,8,8-Heptamethylnonane, clear, colourless liquid
2,2,4,4,6,8,8-Heptamethylnonane

2,2,4,4,6,8,8-Heptamethylnonane was used as the carrier phase to avoid toxic effects of the pure substances. It is also used as a detergent cleansing agent, emulsifier, and thickening agent in cosmetics.

Vzorec: C₁₆H₃₄
MW: 226.45
MDL Number: MFCD00008856
CAS číslo: 4390-04-9
UN: 3295
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR