Tisk…

2,6-Di-O-methyl-β-cyclodextrine, white powder

Dodavatel: MP Biomedicals

Synonyms: Dimethyl-β-cyclodextrin, methylated β-cyclodextrins, Heptakis(2,6-di-o-methyl)-β-cyclodextrin, 2,6-Di-O-methyl-β-cyclodextrin

0215732001 0215732005
ICNA0215732001EA 3900 CZK
ICNA0215732001 ICNA0215732005
2,6-Di-O-methyl-β-cyclodextrine, white powder
2,6-Di-O-methyl-β-cyclodextrine

Heptakis(2,6-di-O-methyl)-β-cyclodextrin is a cholesterol sequestering agent.


Heptakis(2,6-Di-o-methyl)-beta-cyclodexrin is used as reactant involved in Physicochemical studies of inclusion complexes, Probing diffusion and single molecule interactions with reconstituted membrane proteins, Interactions with micelles causing micellar rupture. In Studying cellular transport, used as a cell penetration enhancer,Physicochemical and biopharmaceutical improvement of drugs particularly with solubilization and stability, Interactions with cholesterol.


Soluble in water and ethanol.

Vzorec: C₅₆H₉₈O₃₅
MW: 1331,3563 g/mol
MDL Number: MFCD00011616
CAS číslo: 51166-71-3

Objednat

Výsledky specifikačního testu

Identity Test Passes
FTIR Conforms to Reference
Melting Point Record Result

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR