Tisk…

Palmitoleic acid ≥99%

Dodavatel: MP Biomedicals
Varování

Synonyms: (Z)-Hexadec-9-enoic acid

0215604480 0215604490 0215604491
ICNA0215604480EA 2040 CZK
ICNA0215604480 ICNA0215604490 ICNA0215604491
Palmitoleic acid ≥99%
Palmitoleic acid

Palmitoleic acid is a 16-carbon, omega-7, monounsaturated fatty acid that is enriched in the triglycerides of human adipose tissue and in liver.


Palmitoleic acid (C16:1 n-7) was shown in diabetic mice to attenuate hyperglycemia and hypertriglyceridemia by increasing insulin sensitivity, in part owing to suppression of pro-inflammatory gene expressions and improving hepatic lipid metabolism. It was shown to possibly influence fatty liver deposition/production, insulin action, palmitate, and fatty acid synthase, leading to proposal of a new term, "lipokine" having hormone-like effects.

Vzorec: C₁₆H₃₀O₂
MW: 254.41
Bod tání: –0.1 °C
Storage Temperature: Freezer
MDL Number: MFCD00004437
CAS číslo: 373-49-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR