Tisk…

Morin, light brown powder C.I. 75660

Dodavatel: MP Biomedicals
Varování

Synonyms: 2',3,4',5,7-Pentahydroxy-flavone, C.I. 75660, 2-(2,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one, 2′,3,4′,5,7-Pentahydroxyflavone, Aluminum Ionophore I

0215573305 0215573310
ICNA0215573305EA 1700 CZK
ICNA0215573305 ICNA0215573310
Morin, light brown powder C.I. 75660
Morin

Morin is a flavonoid with anti-oxidant properties.


It has been shown to protect cells against oxygen radical damage. Morin not only scavenges oxyradicals, but also moderately inhibits xanthine oxidase, a free-radical generating enzyme. At concentrations of 75-100 micromolar, morin inhibits oxidation of low density lipoprotein (LDL) by free radicals or Cu2+. Morin has been reported to inhibit rat brain phospha­tidyl­inositol­phos­phate kinase activity in vitro and in vivo.


Soluble in ethanol, methanol (0,5% w/v- clear yellow to brown solution) It is also soluble in water (0,25 mg/ml, 20 °C; 0,94 mg/ml, 100 °C), freely soluble in alcohol, slightly soluble in ether and acetic acid, and soluble in aqueous alkaline solutions.

Vzorec: C₁₅H₁₀O₇
MW: 302,24 g/mol
Bod tání: 303…304 °C
Storage Temperature: Ambient
MDL Number: MFCD00006826
CAS číslo: 480-16-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR