Tisk…

3-(Trimethoxysilyl)propyl methacrylate, clear, colourless liquid

Dodavatel: MP Biomedicals
Varování

Synonyms: γ-methacryloxypropyl-trimethoxysilane, 3-(Methacryloyloxy)propyltrimethoxysilane

0215537805 0215537825
ICNA0215537805EA 1540 CZK
ICNA0215537805 ICNA0215537825
3-(Trimethoxysilyl)propyl methacrylate, clear, colourless liquid
3-(Trimethoxysilyl)propyl methacrylate

Adhesive


γ-Methacryloxypropyl-trimethoxysilane is used in organometallic reagents, organosilicon and trialkoxysilanes. 3-(Trimethoxysilyl)propyl methacrylate has been used to develop an effective method for synthesizing magnetite/polymer colloidal composite microspheres with controllable variations in size and shape of the nanostructures and desirable interfacial chemical functionalities. 3-(Trimethoxysilyl)propyl methacrylate has also been used to carry out the synthesis of hybrid materials possessing zinc oxide nanoparticles encapsulated in core-shell polymer particles having poly(methyl methacrylate) core and chitosan shell (PMMA-CS/ZnO) via an emulsifier-free emulsion polymerization.


Insoluble in water, reacts with water.

Vzorec: C₁₀H₂₀O₅Si
MW: 248.35 g/mol
Bod varu: 190 °C
Bod vzplanutí: 92 °C (ot. pohárek)
Storage Temperature: Refrigerator
MDL Number: MFCD00008593
CAS číslo: 2530-85-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR