Tisk…

Levamisole hydrochloride, white crystalline powder

Dodavatel: MP Biomedicals
Nebezpečí

0215522805 0215522810
ICNA0215522805EA 1810 CZK
ICNA0215522805 ICNA0215522810
Levamisole hydrochloride, white crystalline powder
Levamisole hydrochloride

Levamisole hydrochloride is suitable as an inhibitor of various mammalian alkaline phosphatases.


  • Intestinal alkaline phsophatases are only slightly inhibited
  • Biological response modifier with anthelmintic activity
  • Stereospecific inhibitor of succinic dehydrogenase.


Shows both immunostimulant and immunosuppressant effects, depending on several controllable factors. Very effective in treatment of ascariasis (hookworm infestation). Useful in chemotherapy of colorectal cancers, possibly due to its stimulation of IL-1 production and direct activation of macrophages.


Shows both immunostimulant and immunosuppressant effects, depending on several controllable factors. Very effective in treatment of ascariasis (hookworm infestation). Useful in chemotherapy of colorectal cancers, possibly due to its stimulation of IL-1 production and direct activation of macrophages.

Vzorec: C₁₁H₁₂N₂S·HCl
MDL Number: MFCD00012675
CAS číslo: 16595-80-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR