Tisk…

Imidazolidinyl urea, white powder

Dodavatel: MP Biomedicals
Varování

0215500980 0215500983
ICNA0215500980EA 2840 CZK
ICNA0215500980 ICNA0215500983
Imidazolidinyl urea, white powder
Imidazolidinyl urea

Imidazolidinyl urea is an antimicrobial preservative.


It is topical formulas for use as antioxidants It is used in various studies such as Biological studies investigating the antibacterial synergistic activity of essential oils and surfactants against P. aeruginosa and S. aureus, Pharmacological studies investigating nutrient-sensitized screening for drugs that shift energy metabolism from mitochondrial respiration to glycolysis, Therapeutic studies involving nanostructured vegetable-based carriers for topical delivery of active molecules, In comparison studies of MEKC and microemulsion electrokinetic chromatography for analytical studies of preservatives, Investigations of in vitro induction of apoptosis versus necrosis.


Solubility : Soluble in water(50 mg/ml).


Upozornení: Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.

Vzorec: C₁₁H₁₆N₈O₈
Storage Temperature: Refrigerator
MDL Number: MFCD00221482
CAS číslo: 39236-46-9

Objednat

Výsledky specifikačního testu

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR