Tisk…

Olanzapine ≥98%, crystalline solid

Dodavatel: Cayman Chemical

11937-25 11937-50 11937-100
CAYM11937-25EA 1320 CZK
CAYM11937-25 CAYM11937-50 CAYM11937-100
Olanzapine ≥98%, crystalline solid
Olanzapine

Olanzapine is an antipsychotic, thienobenzodiazepine compound that has been approved for use in the treatment of schizophrenia and bipolar disorder (formulated for human use under the name Zyprexa®).


Olanzapine binds as an antagonist/inverse agonist to dopamine D1 (Ki = 31 nM), dopamine D2 (Ki = 11 nM), dopamine D4 (Ki = 27 nM), serotonin 5-HT2A (Ki = 4 nM), serotonin 5-HT2C (Ki = 11 nM), serotonin 5-HT3 (Ki = 57 nM), muscarinic M1 (Ki = 2 nM), α1-adrenergic (Ki = 19 nM), and histamine H1 (Ki = 7 nM) receptors.


Olanzapine has a high affinity for dopamine D1 and D2 receptors (IC50s = 25 and 10 nM, respectively) in human striatal tissue and for 5-HT2A, 5-HT2C, muscarinic M1, and α1-adrenergic receptors (IC50s = 7, 71, 2, and 70 nM, respectively) in human frontal cortex.

Vzorec: C₁₇H₂₀N₄S
MDL Number: MFCD00866702
CAS číslo: 132539-06-1

Objednat

Výsledky specifikačního testu

Purity ≥98%
Stability ≥2 years at −20 °C
Form Crystalline Solid
UV/Vis λmax: 226, 272 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR